4-Chlor-o-phenylendiamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10823.14
A10823.22
A10823.36
A10823.14EA
66.3
EUR
A10823.14
A10823.22
A10823.36
4-Chlor-o-phenylendiamin 97%
4-Chlor-o-phenylendiamin
Purity: 97%
CAS No.: 95-83-0
Molecular Formula: C6H7ClN2
Formula Weight: 142.59
Melting Point: 70-74°
MDL No.: MFCD00011691
Formel:
C₆H₇ClN₂ Siedepunkt: 280 °C (1013 hPa) Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011691 CAS-Nummer: 95-83-0 EINECS: 202-456-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...