4'-Hydroxychalkon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4'-Hydroxybenzylidenacetophenon

A10838.06 A10838.14
A10838.06EA 40.5 EUR
A10838.06 A10838.14
4'-Hydroxychalkon ≥97%
4'-Hydroxychalkon

Purity: 97% CAS No.: 2657-25-2 Molecular Formula: C15H12O2 Formula Weight: 224.26 Melting Point: 175-177° Beilstein Reference: 2049150

Formel: C₁₅H₁₂O₂
Molecular Weight: 224.26 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016484
CAS-Nummer: 2657-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR