3-Ethyl-2-iodtoluol ≥98%

Lieferant: Thermo Fisher Scientific

A10839.06 A10839.14
A10839.06EA 49.8 EUR
A10839.06 A10839.14
3-Ethyl-2-iodtoluol ≥98%
3-Ethyl-2-iodtoluol

Purity: 98% CAS No.: 175277-95-9 Molecular Formula: C9H11I Formula Weight: 246.08 Boiling Point: 118-120°/15mm MDL No.: MFCD00052855

Formel: C₉H₁₁I
Siedepunkt: 118…120 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052855
CAS-Nummer: 175277-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR