TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)

Lieferant: PanReac AppliChem
Achtung

Synonyme: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

A1084.1000
A1084.1000EA 1040 EUR
A1084.1000
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
TES (2-(Tris(hydroxymethyl)methylamino)ethansulfonsäure)
Formel: C₆H₁₅NO₆S
Molecular Weight: 229,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007532
CAS-Nummer: 7365-44-8
EINECS: 230-906-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR