D(+)-Mannose 99

Lieferant: Thermo Fisher Scientific

Synonyme: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

A10842.14 A10842.22 A10842.36
A10842.14EA 45.7 EUR
A10842.14 A10842.22 A10842.36
D(+)-Mannose 99
D(+)-Mannose

Purity: 99% CAS No.: 3458-28-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 127-133° Solubility: Soluble in water Beilstein Reference: 1564373 Merck Reference: 14,5747 Optical Rotation: +13.8° (c=10 in water, 24h) MDL No.: MFCD00064122

Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Siedepunkt: 376
Schmelzpunkt: 127…133 °C
Dichte: 1.54
Lagertemperatur: Raumtemperatur
MDL: MFCD00064122
CAS-Nummer: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR