Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) ≥99,9%, ultrarein
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyme:
Aminomethylidintrimethanol, Tris(hydroxymethyl)methylamin, THAM, 2-Amino-2-(hydroxymethyl)propan-1,3-diol, Tromethamin, TRIS Base, 2-Amino-2-(hydroxymethyl)-1,3-propandiol
A1086.1000
A1086.5000
A1086.9010
A1086.1000EA
97.3
EUR
A1086.1000
A1086.5000
A1086.9010
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol) ≥99,9%, ultrarein
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel:
NH₂C(CH₂OH)₃ Molecular Weight: 121.14 g/mol Siedepunkt: 357 Schmelzpunkt: 169…172 °C Dichte: 1.328 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004679 CAS-Nummer: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Spezifikation Testergebnisse
Assay (titr.) | min. 99.9 % |
pH (1 M; H2O; 20°C) | 10.5 - 11.5 |
Water | max. 0.2 % |
Cu | max. 0.0001 % |
Fe | max. 0.0001 % |
PB | max. 0.0001 % |
A (1 cm/1 M in H2O) at 430 nm | max. 0.004 |
A (1 cm/1 M in H2O) at 260 nm | 0.03 |
A (1 cm/1 M in H2O) at 280 nm | 0.02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...