2,6-Dichlorbenzamid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
2,6-Dichlorbenzoesäureamid
A10865.06
A10865.14
A10865.22
A10865.06EA
24
EUR
A10865.06
A10865.14
A10865.22
2,6-Dichlorbenzamid ≥98%
2,6-Dichlorbenzamid
Purity: 98%
CAS No.: 2008-58-4
Molecular Formula: C7H5Cl2NO
Formula Weight: 190.03
Melting Point: 198-200°
Beilstein Reference: 1869103
Formel:
C₇H₅Cl₂NO Siedepunkt: 335 °C (1013 hPa) Schmelzpunkt: 198…200 °C Dichte: 1,44 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007975 CAS-Nummer: 2008-58-4 EINECS: 217-918-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...