TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)

Lieferant: PanReac AppliChem

A1087.0500 A1087.1000 A1087.5000
A1087.0500EA 111 EUR
A1087.0500 A1087.1000 A1087.5000
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
Formel: NH₂C(CH₂OH)₃·HCl
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012590
CAS-Nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR