Pyridiniumchlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A10871.0B
A10871.18
A10871.30
A10871.0BEA
289
EUR
A10871.0B
A10871.18
A10871.30
Pyridiniumchlorid ≥98%
Pyridinium chloride
Purity: 98%
CAS No.: 628-13-7
Molecular Formula: C5H5N.HCl
Formula Weight: 115.56
Melting Point: ca 145°
Boiling Point: 222-224°
MDL No.: MFCD00012802
Formel:
C₅H₆ClN Siedepunkt: 222…224 °C Schmelzpunkt: ∼ 145 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012802 CAS-Nummer: 628-13-7 EINECS: 211-027-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...