Methansulfonamid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methylsulphonamide

A10885.09 A10885.18 A10885.30 A10885.36
A10885.09EA 31 EUR
A10885.09 A10885.18 A10885.30 A10885.36
Methansulfonamid ≥98%
Methansulfonamid

Purity: 98+% CAS No.: 3144-09-0 Molecular Formula: CH5NO2S Formula Weight: 95.13 Melting Point: 87-93° Beilstein Reference: 1740835

Formel: CH₅NO₂S
Schmelzpunkt: 87…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007940
CAS-Nummer: 3144-09-0
EINECS: 221-553-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR