Methansulfonamid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methylsulphonamide
A10885.09
A10885.18
A10885.30
A10885.36
A10885.09EA
31
EUR
A10885.09
A10885.18
A10885.30
A10885.36
Methansulfonamid ≥98%
Methansulfonamid
Purity: 98+%
CAS No.: 3144-09-0
Molecular Formula: CH5NO2S
Formula Weight: 95.13
Melting Point: 87-93°
Beilstein Reference: 1740835
Formel:
CH₅NO₂S Schmelzpunkt: 87…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007940 CAS-Nummer: 3144-09-0 EINECS: 221-553-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...