2'-Aminoacetophenon 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
o-Aminoacetophenon
A10895.06
A10895.18
A10895.30
A10895.09
A10895.06EA
37.9
EUR
A10895.06
A10895.18
A10895.30
A10895.09
2'-Aminoacetophenon 98%
2'-Aminoacetophenon
Purity: 98%
CAS No.: 551-93-9
Molecular Formula: C8H9NO
Formula Weight: 135.17
Storage Temperature: 2°C to 8°C
Flash Point: 112°(233°F)
Refractive Index: 1.6160
Boiling Point: 253-255°
Beilstein Reference: 386122
Merck Reference: 14,413
MDL No.: MFCD00007717
Formel:
C₈H₉NO Molecular Weight: 135.17 g/mol Siedepunkt: 253…255 °C Schmelzpunkt: 20 Dichte: 1.118 Flammpunkt: 112 °C (233 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00007717 CAS-Nummer: 551-93-9 EINECS: 209-002-8 Merck Index: 13,00412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...