2'-Aminoacetophenon 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: o-Aminoacetophenon

A10895.06 A10895.18 A10895.30 A10895.09
A10895.06EA 37.9 EUR
A10895.06 A10895.18 A10895.30 A10895.09
2'-Aminoacetophenon 98%
2'-Aminoacetophenon

Purity: 98% CAS No.: 551-93-9 Molecular Formula: C8H9NO Formula Weight: 135.17 Storage Temperature: 2°C to 8°C Flash Point: 112°(233°F) Refractive Index: 1.6160 Boiling Point: 253-255° Beilstein Reference: 386122 Merck Reference: 14,413 MDL No.: MFCD00007717

Formel: C₈H₉NO
Molecular Weight: 135.17 g/mol
Siedepunkt: 253…255 °C
Schmelzpunkt: 20
Dichte: 1.118
Flammpunkt: 112 °C (233 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00007717
CAS-Nummer: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR