3-Brombenzylalkohol ≥99%

Lieferant: Thermo Scientific

Synonyme: (3-Bromophenyl)methanol

A10929.06
A10929.06EA 44.7 EUR
A10929.06
3-Brombenzylalkohol ≥99%
3-Brombenzylalkohol

Purity: 99% CAS No.: 15852-73-0 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 110°(230°F) Refractive Index: 1.5840 Boiling Point: 165°/16mm Beilstein Reference: 2242512 MDL No.: MFCD00004629

Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 165 °C (16 mmHg)
Dichte: 1,560
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004629
CAS-Nummer: 15852-73-0
EINECS: 239-975-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR