2,4-Dichlorphenylhydraziniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(2,4-Dichlorphenyl)hydrazin Hydrochlorid, 2,4-Dichlorphenylhydrazinhydrochlorid, 2,4-Dichlorphenylhydrazin Hydrochlorid

A10943.09 A10943.18 A10943.30
A10943.09EA 67 EUR
A10943.09 A10943.18 A10943.30
2,4-Dichlorphenylhydraziniumchlorid ≥98%
2,4-Dichlorphenylhydraziniumchlorid

Purity: 98% CAS No.: 5446-18-4 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.49 Melting Point: ca 212° dec. Beilstein Reference: 3708828

Formel: C₆H₇Cl₃N₂
Molecular Weight: 213.49 g/mol
Schmelzpunkt: ∼ 212 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012929
CAS-Nummer: 5446-18-4
EINECS: 226-659-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR