Ethyl-3,4-dihydroxybenzoat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Protocatechusäureethylester, 3,4-Dihydroxybenzoesäureethylester

A11001.09 A11001.18 A11001.30 AB111834.0050
A11001.09EA 52.2 EUR
A11001.09 A11001.18 A11001.30 ABCRAB111834.0050
Ethyl-3,4-dihydroxybenzoat ≥98%
Ethyl-3,4-dihydroxybenzoat

Purity: 98% CAS No.: 3943-89-3 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 132-135° Beilstein Reference: 2097435

Formel: C₉H₁₀O₄
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002199
CAS-Nummer: 3943-89-3
EINECS: 223-529-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR