Dimethylmalonat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester

A11007.22 A11007.0E A11007.36
A11007.22EA 24.2 EUR
A11007.22 A11007.0E A11007.36
Dimethylmalonat ≥98%
Dimethylmalonat

Purity: 98+% CAS No.: 108-59-8 Molecular Formula: C5H8O4 Formula Weight: 132.12 Flash Point: 90°(194°F) Refractive Index: 1.4130 Melting Point: -62° Boiling Point: 180-181° Beilstein Reference: 774261 Merck Reference: 14,6096 MDL No.: MFCD00008460

Formel: C₅H₈O₄
Molecular Weight: 132.12 g/mol
Siedepunkt: 180…181 °C
Schmelzpunkt: –62 °C
Dichte: 1.156
Flammpunkt: 90 °C (194 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008460
CAS-Nummer: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR