3-Nitro-L-tyrosin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine

A11018.06 A11018.14 A11018.22
A11018.06EA 57 EUR
A11018.06 A11018.14 A11018.22
3-Nitro-L-tyrosin ≥98%
3-Nitro-L-tyrosin

Purity: 98% CAS No.: 621-44-3 Molecular Formula: C9H10N2O5 Formula Weight: 226.19 Melting Point: ca 230° dec. Beilstein Reference: 2813157

Formel: C₉H₁₀N₂O₅
Molecular Weight: 226.3 g/mol
Schmelzpunkt: ∼ 230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007123
CAS-Nummer: 621-44-3
EINECS: 210-688-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR