4'-Ethoxyacetophenon 98
Lieferant: Thermo Fisher Scientific
A11031.06
A11031.14
A11031.22
A11031.06EA
29.3
EUR
A11031.06
A11031.14
A11031.22
4'-Ethoxyacetophenon 98
4'-Ethoxyacetophenon
Purity: 98%
CAS No.: 1676-63-7
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 134°(273°F)
Melting Point: 35-38°
Boiling Point: 145-146°/13mm
Beilstein Reference: 636783
MDL No.: MFCD00009095
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164.2 g/mol Siedepunkt: 145…146 °C (13 mmHg) Schmelzpunkt: 35…38 °C Flammpunkt: 134 °C (273 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009095 CAS-Nummer: 1676-63-7 EINECS: 216-825-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...