4'-Ethoxyacetophenon 98

Lieferant: Thermo Fisher Scientific

A11031.06 A11031.14 A11031.22
A11031.06EA 29.3 EUR
A11031.06 A11031.14 A11031.22
4'-Ethoxyacetophenon 98
4'-Ethoxyacetophenon

Purity: 98% CAS No.: 1676-63-7 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 134°(273°F) Melting Point: 35-38° Boiling Point: 145-146°/13mm Beilstein Reference: 636783 MDL No.: MFCD00009095

Formel: C₁₀H₁₂O₂
Molecular Weight: 164.2 g/mol
Siedepunkt: 145…146 °C (13 mmHg)
Schmelzpunkt: 35…38 °C
Flammpunkt: 134 °C (273 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009095
CAS-Nummer: 1676-63-7
EINECS: 216-825-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR