1-Naphthylessigsäure 96% may cont. up to 4% 2-isomer

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: α-Naphthylessigsäure, Planofix, 1-Naphthalinessigsäure

A11069.0B A11069.18 A11069.30
A11069.0BEA 284 EUR
A11069.0B A11069.18 A11069.30
1-Naphthylessigsäure 96% may cont. up to 4% 2-isomer
1-Naphthylessigsäure

Purity: 96% CAS No.: 86-87-3 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 126-133° Beilstein Reference: 1308415 Merck Reference: 14,6371 MDL No.: MFCD00004046

Formel: C₁₂H₁₀O₂
Molecular Weight: 186.21 g/mol
Siedepunkt: 352
Schmelzpunkt: 126…133 °C
Dichte: ∼1.263 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004046
CAS-Nummer: 86-87-3
EINECS: 201-705-8
Merck Index: 12,06458

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR