Natriumbutyrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Buttersäure Natriumsalz

A11079.06 A11079.22 A11079.36
A11079.06EA 28.7 EUR
A11079.06 A11079.22 A11079.36
Natriumbutyrat ≥98%
Natriumbutyrat

Purity: 98+% CAS No.: 156-54-7 Molecular Formula: C4H7NaO2 Formula Weight: 110.09 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 300° Beilstein Reference: 3629439 MDL No.: MFCD00002816


Decreases calcium release from intracellular stores. Inhibits histone deacetylase

Formel: C₄H₇NaO₂
Molecular Weight: 110.09 g/mol
Schmelzpunkt: >300 °C
Dichte: 1.324 g/cm³ (30 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002816
CAS-Nummer: 156-54-7
EINECS: 205-857-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR