1,4-Cyclohexandion ≥98%

Lieferant: Thermo Fisher Scientific

A11100.14 A11100.22 A11100.36
A11100.14EA 62.5 EUR
A11100.14 A11100.22 A11100.36
1,4-Cyclohexandion ≥98%
1,4-Cyclohexandion

Purity: 98% CAS No.: 637-88-7 Molecular Formula: C6H8O2 Formula Weight: 112.13 Storage Temperature: 2°C to 8°C Flash Point: 132°(269°F) Melting Point: 77-78° Boiling Point: 132°/20mm Beilstein Reference: 774152 MDL No.: MFCD00001606

Formel: C₆H₈O₂
Molecular Weight: 112.13 g/mol
Siedepunkt: 132 °C (20 mmHg)
Schmelzpunkt: 77…78 °C
Dichte: 1.28 g/cm³ (20 °C)
Flammpunkt: 132 °C (269 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00001606
CAS-Nummer: 637-88-7
EINECS: 211-306-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR