2-Brom-5-fluortoluol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Bromo-4-fluoro-2-methylbenzene

A11112.14 A11112.22 A11112.06
A11112.14EA 57.8 EUR
A11112.14 A11112.22 A11112.06
2-Brom-5-fluortoluol ≥98%
2-Brom-5-fluortoluol

Purity: 98+% CAS No.: 452-63-1 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 70°(158°F) Refractive Index: 1.5260 Boiling Point: 169-170° Beilstein Reference: 1859121 MDL No.: MFCD00017921

Formel: C₇H₆BrF
Molecular Weight: 189.03 g/mol
Siedepunkt: 169…170 °C
Dichte: 1.495
Flammpunkt: 70 °C (158 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017921
CAS-Nummer: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR