Ethylformiat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11113.0F
A11113.AE
A11113.AP
A11113.0FEA
136
EUR
A11113.0F
A11113.AE
A11113.AP
Ethylformiat ≥97%
Ethylformiat
Purity: 97%
CAS No.: 109-94-4
Molecular Formula: C3H6O2
Formula Weight: 74.08
Storage Temperature: 2°C to 8°C
Flash Point: -19°(-2°F)
Physical Form: Liquid
Density: 0.921
Refractive Index: 1.3600
Appearance: Colourless
Odour: Fruit-like
pH: 4.1
Melting Point: -81°
Boiling Point: 52-54°
Beilstein Reference: 906769
Merck Reference: 14,3807
UN No.: 1190
MDL No.: MFCD00003294
Formel:
C₃H₆O₂ Siedepunkt: 52…54 °C Schmelzpunkt: –81 °C Dichte: 0,921 Flammpunkt: –19 °C (–2 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00003294 CAS-Nummer: 109-94-4 EINECS: 203-721-0 UN: 1190 ADR: 3,II Merck Index: 14,3807 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...