L(-)-Tyrosin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

A11141.0B A11141.18 A11141.22 A11141.30
A11141.0BEA 294 EUR
A11141.0B A11141.18 A11141.22 A11141.30
L(-)-Tyrosin ≥99%
L(-)-Tyrosin

Purity: 99% CAS No.: 60-18-4 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Physical Form: Cryst powder Density: 1.46 Appearance: White Melting Point: 300° Beilstein Reference: 392441 Merck Reference: 14,9839 Optical Rotation: -11° (c=4 in 1N HCl) MDL No.: MFCD00002606

Formel: C₉H₁₁NO₃
Siedepunkt: 351 °C (1013 hPa)
Schmelzpunkt: >300 °C
Dichte: 1,46
Lagertemperatur: Raumtemperatur
MDL: MFCD00002606
CAS-Nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 14,9839

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR