1-Octen ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11146.0F
A11146.AP
A11146.0FEA
113
EUR
A11146.0F
A11146.AP
1-Octen ≥97%
1-Octen
Purity: 97+%
CAS No.: 111-66-0
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 21°(70°F)
Refractive Index: 1.4090
Odour: Petroleum-like
Melting Point: -102°
Boiling Point: 122-123°
Beilstein Reference: 1734497
Merck Reference: 14,1764
UN No.: 3295
MDL No.: MFCD00009548
Formel:
C₈H₁₆ Siedepunkt: 122…123 °C Schmelzpunkt: –102 °C Dichte: 0,715 Flammpunkt: 21 °C (70 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009548 CAS-Nummer: 111-66-0 EINECS: 203-893-7 UN: 3295 ADR: 3,II Merck Index: 14,1764 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...