Formamidinacetat 99

Lieferant: Thermo Fisher Scientific
Achtung

A11158.22 A11158.36
A11158.22EA 51.5 EUR
A11158.22 A11158.36
Formamidinacetat 99
Formamidinacetat

Purity: 99% CAS No.: 3473-63-0 Molecular Formula: C3H8N2O2 Formula Weight: 104.11 pH: 8 Melting Point: ca 158° dec. Beilstein Reference: 3563488 MDL No.: MFCD00012866

Formel: C₃H₈N₂O₂
Molecular Weight: 104.11 g/mol
Schmelzpunkt: ∼ 158 °C (Zers.)
Dichte: 1.124
Lagertemperatur: Raumtemperatur
MDL: MFCD00012866
CAS-Nummer: 3473-63-0
EINECS: 222-442-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR