Vanillin ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11169.0E
A11169.22
A11169.36
A11169.0EEA
458
EUR
A11169.0E
A11169.22
A11169.36
Vanillin ≥99%
Vanillin
Purity: 99%
CAS No.: 121-33-5
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point: 153°(307°F)
Physical Form: Crystalline powder
Density: 1.06
Sensitivity: Air and Light Sensitive
Appearance: White
Melting Point: 81-83°
Boiling Point: 170°/15mm
Beilstein Reference: 472792
Merck Reference: 14,9932
MDL No.: MFCD00006942
Formel:
C₈H₈O₃ Siedepunkt: 170 °C (15 mmHg) Schmelzpunkt: 81…83 °C Dichte: 1,06 Flammpunkt: 153 °C (307 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006942 CAS-Nummer: 121-33-5 EINECS: 204-465-2 Merck Index: 14,9932 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...