Benzonitril ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 100-47-0
Molecular Formula: C7H5N
Formula Weight: 103.12
Flash Point: 71°(159°F)
Refractive Index: 1.5285
Melting Point: -13°
Boiling Point: 191-193°
Beilstein Reference: 506893
Merck Reference: 14,1097
UN No.: 2224
MDL No.: MFCD00001770
Formel:
C₇H₅N Siedepunkt: 191…193 °C Schmelzpunkt: –13 °C Dichte: 1,007 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001770 CAS-Nummer: 100-47-0 EINECS: 202-855-7 UN: 2224 ADR: 6.1,II Merck Index: 14,1097 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...