Natriumdodecylsulfat (SDS), (max. 1,5% H₂O) 99% (Trockengewicht)

Lieferant: Thermo Fisher Scientific
Gefahr

A11183.14 A11183.18 A11183.22 A11183.30 A11183.36
A11183.14EA 34.9 EUR
A11183.14 A11183.18 A11183.22 A11183.30 A11183.36
Natriumdodecylsulfat (SDS), (max. 1,5% H₂O) 99% (Trockengewicht)
Natriumdodecylsulfat (SDS)

Purity: 99% CAS No.: 151-21-3 Molecular Formula: CH3(CH2)11OSO3Na Formula Weight: 288.38 Physical Form: Powder Appearance: White Melting Point: ca 206° Beilstein Reference: 3599286 Merck Reference: 14,8636 UN No.: 2926 MDL No.: MFCD00036175


Denatures and solubilizes proteins for electrophoresis

Formel: CH₃(CH₂)₁₁OSO₃Na
Schmelzpunkt: ∼206 °C
Dichte: 1,1 g/cm³ (20 °C)
Flammpunkt: >100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036175
CAS-Nummer: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR