5-Brom-1-penten 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11219.06
A11219.14
A11219.06EA
76.8
EUR
A11219.06
A11219.14
5-Brom-1-penten 96%
5-Brom-1-penten
Purity: 96%
CAS No.: 1119-51-3
Molecular Formula: BrC5H9
Formula Weight: 149.03
Storage Temperature: 2°C to 8°C
Flash Point: 30°(86°F)
Refractive Index: 1.4639
Boiling Point: 126-127°
Beilstein Reference: 506077
UN No.: 1993
MDL No.: MFCD0000-364
Formel:
C₅H₉Br Siedepunkt: 126…127 °C Dichte: 1,259 Flammpunkt: 30 °C (86 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000264 CAS-Nummer: 1119-51-3 EINECS: 214-281-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...