5-Brom-1-penten 96%

Lieferant: Thermo Fisher Scientific
Achtung

A11219.06 A11219.14
A11219.06EA 76.8 EUR
A11219.06 A11219.14
5-Brom-1-penten 96%
5-Brom-1-penten

Purity: 96% CAS No.: 1119-51-3 Molecular Formula: BrC5H9 Formula Weight: 149.03 Storage Temperature: 2°C to 8°C Flash Point: 30°(86°F) Refractive Index: 1.4639 Boiling Point: 126-127° Beilstein Reference: 506077 UN No.: 1993 MDL No.: MFCD0000-364

Formel: C₅H₉Br
Siedepunkt: 126…127 °C
Dichte: 1,259
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00000264
CAS-Nummer: 1119-51-3
EINECS: 214-281-4
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR