(1H-Benzoimidazol-2-ylsulphanyl)acetic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-((1H-Benzo[d]imidazol-2-yl)thio)acetic acid

A11221.06 A11221.14
A11221.06EA 56.1 EUR
A11221.06 A11221.14
(1H-Benzoimidazol-2-ylsulphanyl)acetic acid 98%
(1H-Benzoimidazol-2-ylsulphanyl)acetic acid

Purity: 98% CAS No.: 3042-00-0 Molecular Formula: C9H8N2O2S Formula Weight: 208.24 Melting Point: ca 215° dec. Beilstein Reference: 11825 MDL No.: MFCD00022673

Formel: C₉H₈N₂O₂S
Schmelzpunkt: ∼ 215 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00022673
CAS-Nummer: 3042-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR