(1H-Benzoimidazol-2-ylsulphanyl)acetic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-((1H-Benzo[d]imidazol-2-yl)thio)acetic acid
A11221.06
A11221.14
A11221.06EA
56.1
EUR
A11221.06
A11221.14
(1H-Benzoimidazol-2-ylsulphanyl)acetic acid 98%
(1H-Benzoimidazol-2-ylsulphanyl)acetic acid
Purity: 98%
CAS No.: 3042-00-0
Molecular Formula: C9H8N2O2S
Formula Weight: 208.24
Melting Point: ca 215° dec.
Beilstein Reference: 11825
MDL No.: MFCD00022673
Formel:
C₉H₈N₂O₂S Schmelzpunkt: ∼ 215 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022673 CAS-Nummer: 3042-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...