5-Chlor-2-nitroanilin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A11241.06 A11241.14
A11241.06EA 32.1 EUR
A11241.06 A11241.14
5-Chlor-2-nitroanilin ≥97%
5-Chlor-2-nitroanilin

Purity: 97% CAS No.: 1635-61-6 Molecular Formula: C6H5ClN2O2 Formula Weight: 172.57 Odour: Weak, characteristic Melting Point: 124-128° Beilstein Reference: 2210201 UN No.: 2237 MDL No.: MFCD00007776

Formel: C₆H₅ClN₂O₂
Siedepunkt: 323 °C (1013 hPa)
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007776
CAS-Nummer: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR