5-Chlor-2-nitroanilin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11241.06
A11241.14
A11241.06EA
32.1
EUR
A11241.06
A11241.14
5-Chlor-2-nitroanilin ≥97%
5-Chlor-2-nitroanilin
Purity: 97%
CAS No.: 1635-61-6
Molecular Formula: C6H5ClN2O2
Formula Weight: 172.57
Odour: Weak, characteristic
Melting Point: 124-128°
Beilstein Reference: 2210201
UN No.: 2237
MDL No.: MFCD00007776
Formel:
C₆H₅ClN₂O₂ Siedepunkt: 323 °C (1013 hPa) Schmelzpunkt: 124…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007776 CAS-Nummer: 1635-61-6 EINECS: 216-661-5 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...