Natriumperchlorat Monohydrat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A11243.0B A11243.0I A11243.30 A11243.C4
A11243.0BEA 141 EUR
A11243.0B A11243.0I A11243.30 A11243.C4
Natriumperchlorat Monohydrat ≥97%
Natriumperchlorat Monohydrat

Purity: 97+% CAS No.: 7791-07-3 Molecular Formula: NaClO4.H2O Formula Weight: 140.48 (122.46anhy) Odour: Odourless Melting Point: 130° dec. UN No.: 1502 MDL No.: MFCD00149164

Formel: NaClO₄·H₂O
Dichte: 2,02
Lagertemperatur: Raumtemperatur
MDL: MFCD00149164
CAS-Nummer: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 14,8653

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR