2-Acetyl-5-methylthiophen ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98%
CAS No.: 13679-74-8
Molecular Formula: C7H8OS
Formula Weight: 140.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5625
Melting Point: 24-28°
Boiling Point: 65-67°/1mm
Beilstein Reference: 110854
MDL No.: MFCD00014529
Formel:
C₇H₈OS Siedepunkt: 65…67 °C (1 mmHg) Schmelzpunkt: 24…28 °C Dichte: 1,119 |
MDL:
MFCD00014529 CAS-Nummer: 13679-74-8 EINECS: 237-181-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...