(3-Aminopropyl)trimethoxysilan 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Trimethoxy(3-aminopropyl)silan, 3-(Trimethoxysilyl)propylamin
A11284.0E
A11284.22
A11284.36
A11284.0EEA
775
EUR
A11284.0E
A11284.22
A11284.36
(3-Aminopropyl)trimethoxysilan 97%
(3-Aminopropyl)trimethoxysilan
Purity: 97%
CAS No.: 13822-56-5
Molecular Formula: C6H17NO3Si
Formula Weight: 179.29
Flash Point: 90°(194°F)
Physical Form: Liquid
Density: 1.02
Refractive Index: 1.4240
Sensitivity: Air and Moisture Sensitive
Appearance: Colourless
Odour: Amine-like
Boiling Point: 194°
Beilstein Reference: 2038118
UN No.: 2735
MDL No.: MFCD00008206
Formel:
H₂NCH₂CH₂CH₂Si(OCH₃)₃ Siedepunkt: 194 °C Dichte: 1,02 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008206 CAS-Nummer: 13822-56-5 EINECS: 237-511-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...