1,1-Diphenylhydrazin Hydrochlorid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A11289.14 A11289.22 A11289.06
A11289.14EA 109 EUR
A11289.14 A11289.22 A11289.06
1,1-Diphenylhydrazin Hydrochlorid ≥98%
1,1-Diphenylhydrazin Hydrochlorid

Purity: 98% CAS No.: 530-47-2 Molecular Formula: C12H12N2.HCl Formula Weight: 220.70 Odour: Odourless Melting Point: ca 163° dec. Beilstein Reference: 3569340 Merck Reference: 14,3325 MDL No.: MFCD00012950

Formel: C₁₂H₁₃ClN₂
Schmelzpunkt: ∼ 163 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012950
CAS-Nummer: 530-47-2
EINECS: 208-481-0
Merck Index: 14,3325

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR