(±)-2-Methylcapronsäure ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(±)-2-Methylhexansäure
A11290.06
A11290.06EA
38.2
EUR
A11290.06
(±)-2-Methylcapronsäure ≥98%
(±)-2-Methylcapronsäure
Purity: 98%
CAS No.: 4536-23-6
Molecular Formula: C7H14O2
Formula Weight: 130.19
Flash Point: 105°(221°F)
Refractive Index: 1.4205
Odour: Rancid
Boiling Point: 209-210°
Beilstein Reference: 1721227
UN No.: 3265
MDL No.: MFCD00002674
Formel:
C₇H₁₄O₂ Molecular Weight: 130,19 g/mol Siedepunkt: 209…210 °C Dichte: 0,916 Flammpunkt: 105 °C (221 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002674 CAS-Nummer: 4536-23-6 EINECS: 224-883-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...