Natriumdihydrogenphosphat Dihydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Natriumphosphat monobasisch Dihydrat
A11316.0E
A11316.36
A11316.22
A11316.0EEA
103
EUR
A11316.0E
A11316.36
A11316.22
Natriumdihydrogenphosphat Dihydrat ≥99%
Natriumdihydrogenphosphat Dihydrat
Purity: 99%
CAS No.: 13472-35-0
Molecular Formula: H2NaO4P.2H2O
Formula Weight: 156.01 (119.98anhy)
Merck Reference: 14,8660
MDL No.: MFCD00149209
Formel:
NaH₂PO₄·2H₂O Siedepunkt: ∼650 °C (1013 hPa) Schmelzpunkt: 60 °C Dichte: 1,910 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149209 CAS-Nummer: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 14,8660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...