1,6-Heptadiin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A11318.06
A11318.14
A11318.22
A11318.06EA
112
EUR
A11318.06
A11318.14
A11318.22
1,6-Heptadiin ≥97%
1,6-Heptadiin
Purity: 97%
CAS No.: 2396-63-6
Molecular Formula: C7H8
Formula Weight: 92.14
Storage Temperature: 2°C to 8°C
Flash Point: 9°(48°F)
Refractive Index: 1.4420
Odour: Recognizable
Boiling Point: 109-111°
Beilstein Reference: 1731756
UN No.: 3295
MDL No.: MFCD00014925
Formel:
C₇H₈ Siedepunkt: 109…111 °C Schmelzpunkt: –85 °C Dichte: 0,810 Flammpunkt: 9 °C (48 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00014925 CAS-Nummer: 2396-63-6 EINECS: 219-253-5 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...