3,4-Dimethoxyphenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A11344.06
A11344.14
A11344.22
A11344.06EA
82.8
EUR
A11344.06
A11344.14
A11344.22
3,4-Dimethoxyphenol ≥98%
3,4-Dimethoxyphenol
Purity: 98%
CAS No.: 2033-89-8
Molecular Formula: C8H10O3
Formula Weight: 154.16
Melting Point: 79-82°
Boiling Point: 167°/14mm
MDL No.: MFCD00008390
Formel:
C₈H₁₀O₃ Siedepunkt: 167 °C (14 mmHg) Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008390 CAS-Nummer: 2033-89-8 EINECS: 217-995-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...