α,α,α,α',α',α'-Hexafluor-m-xylol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
α,α,α,β,β,β-Hexafluor-m-xylol
A11360.0B
A11360.18
A11360.30
A11360.0BEA
312
EUR
A11360.0B
A11360.18
A11360.30
α,α,α,α',α',α'-Hexafluor-m-xylol ≥98%
α,α,α,α',α',α'-Hexafluor-m-xylol
Purity: 98+%
CAS No.: 402-31-3
Molecular Formula: C8H4F6
Formula Weight: 214.11
Flash Point: 26°(78°F)
Refractive Index: 1.3790
Odour: Aromatic
Melting Point: -35°
Boiling Point: 115-117°
Beilstein Reference: 2052589
UN No.: 1993
MDL No.: MFCD00000392
Formel:
C₈H₄F₆ Molecular Weight: 214.11 g/mol Siedepunkt: 115…117 °C Schmelzpunkt: –35 °C Dichte: 1.378 Flammpunkt: 26 °C (78 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000392 CAS-Nummer: 402-31-3 EINECS: 206-939-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...