3,5-Bis(trifluormethyl)phenylboronsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(3,5-Bis(trifluormethyl)phenyl)metaborsäure, 3,5-Bis(trifluormethyl)benzolboronsäure, Dihydroxy(3,5-bis(trifluormethyl)phenyl)boran
A11373.03
A11373.06
A11373.14
A11373.03EA
32.8
EUR
A11373.03
A11373.06
A11373.14
3,5-Bis(trifluormethyl)phenylboronsäure ≥97%
3,5-Bis(trifluormethyl)phenylboronsäure
Purity: 97+%
CAS No.: 73852-19-4
Molecular Formula: C8H5BF6O2
Formula Weight: 257.93
Melting Point: 219-224°
Beilstein Reference: 7379990
Formel:
C₈H₅BF₆O₂ Schmelzpunkt: 219…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051850 CAS-Nummer: 73852-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...