Amidoschwarz 10 B (Naphtholblauschwarz, Säureschwarz 1)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Amino-8-hydroxy-2-((4-nitrophenyl)diazenyl)-7-(phenyldiazenyl)naphthalin-3,6-disulfonsäure Dinatriumsalz

A11374.14 A11374.22
A11374.14EA 36.4 EUR
A11374.14 A11374.22
Amidoschwarz 10 B (Naphtholblauschwarz, Säureschwarz 1)
Amidoschwarz 10 B (Naphtholblauschwarz, Säureschwarz 1)

CAS No.: 1064-48-8 Molecular Formula: C22H14N6Na2O9S2 Formula Weight: 616.49 Odour: Odourless pH: 9.0 Beilstein Reference: 5374263

Formel: C₂₂H₁₄N₆Na₂O₉S₂
Molecular Weight: 616.49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004017
CAS-Nummer: 1064-48-8
EINECS: 213-903-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR