TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid) ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Tris(hydroxymethyl)methylamin Hydrochlorid, 2-Amino-2-(hydroxymethyl)propan-1,3-diolhydrochlorid, 2-Amino-2-(hydroxymethyl)-1,3-propandiol HCl, Tris(hydroxymethyl)methylammoniumchlorid, TRIS HCl, THAM HCl, Trometamol Hydrochlorid
A11379.0B
A11379.18
A11379.30
A11379.0BEA
277
EUR
A11379.0B
A11379.18
A11379.30
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid) ≥99%
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
Purity: 99+%
CAS No.: 1185-53-1
Molecular Formula: C4H11NO3.HCl
Formula Weight: 157.60
pH: 3.5-5.0
Melting Point: ca 152° dec.
Beilstein Reference: 3675235
MDL No.: MFCD00012590
Formel:
NH₂C(CH₂OH)₃·HCl Schmelzpunkt: ∼ 152 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012590 CAS-Nummer: 1185-53-1 EINECS: 214-684-5 Merck Index: 12,09902 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...