3-Chlorbenzamid 98

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Chlorbenzoesäureamid

A11389.09
A11389.09EA 47.8 EUR
A11389.09
3-Chlorbenzamid 98
3-Chlorbenzamid

Purity: 98+% CAS No.: 618-48-4 Molecular Formula: C7H6ClNO Formula Weight: 155.58 Melting Point: 133-136° Beilstein Reference: 1859940

Formel: C₇H₆ClNO
Molecular Weight: 155.58 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014799
CAS-Nummer: 618-48-4
EINECS: 210-554-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR